Tetraethyl orthocarbonate, Sigma-Aldrich®
Dodavatel: Merck
M-Clarity™ quality level = MQ200
163627-25G
163627-5G
SIAL163627-25GEA
6670
CZK
SIAL163627-25G
SIAL163627-5G
Tetraethyl orthocarbonate, Sigma-Aldrich®
Tetraethyl orthocarbonate
Vzorec:
C₉H₂₀O₄ MW: 192,26 g/mol |
MDL Number:
MFCD00009221 CAS číslo: 78-09-1 EINECS: 201-082-2 UN: 3272 ADR: 3,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...