Tisk…

Nicotinic acid hydrazide, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: Nicotinohydrazide, (3-Pyridylcarbonyl)hydrazine, Pyridine-3-carbohydrazide

M-Clarity™ quality level = MQ100

107425-100G 107425-25G
SIAL107425-100GEA 5190 CZK
SIAL107425-100G SIAL107425-25G
Nicotinic acid hydrazide, Sigma-Aldrich®
Nicotinic acid hydrazide
Vzorec: C₆H₇N₃O
MW: 137,14 g/mol
MDL Number: MFCD00006383
CAS číslo: 553-53-7
EINECS: 209-041-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR