Tisk…

Perfluorobutane-1-sulphonyl chloride, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: Nonafluorobutane-1-sulphonyl chloride

51974-1G-F 51974-5G-F
SIAL51974-1G-FEA 3840 CZK
SIAL51974-1G-F SIAL51974-5G-F
Perfluorobutane-1-sulphonyl chloride, Sigma-Aldrich®
Perfluorobutane-1-sulphonyl chloride
Vzorec: C₄ClF₉O₂S
MW: 318,55 g/mol
MDL Number: MFCD03412262
CAS číslo: 2991-84-6
UN: 3265
ADR: 8,II

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR