Ethyl-4,4,4-trifluorobutyrate ≥98%
Dodavatel: Thermo Fisher Scientific
|
Varování
|
Synonyms:
Ethyl 4,4,4-trifluorobutanoate, Ethyl 4,4,4-trifluorobutyrate
L11399.03
L11399.06
L11399.03EA
718
CZK
L11399.03
L11399.06
Ethyl-4,4,4-trifluorobutyrate ≥98%
Ethyl-4,4,4-trifluorobutyrate
Purity: 98%
CAS No.: 371-26-6
Molecular Formula: CF2CH2CH2CO2CH2CH3
Formula Weight: 170.13
Flash Point: 33°(91°F)
Refractive Index: 1.3520
Boiling Point: 126-127°
Beilstein Reference: 1769215
UN No.: 3272
MDL No.: MFCD00041398
Vzorec:
C₆H₉F₃O₂ Bod varu: 126…127 °C Hustota: 1,160 Bod vzplanutí: 33 °C (91 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00041398 CAS číslo: 371-26-6 UN: 3272 ADR: 3,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...