Valeric acid hydrazide 95%
Dodavatel: Thermo Scientific
|
Varování
|
Synonyms:
Pentanehydrazide
L01555.06
L01555.14
L01555.06EA
934
CZK
L01555.06
L01555.14
Valeric acid hydrazide 95%
Valeric acid hydrazide
Purity: 95%
CAS No.: 38291-82-6
Molecular Formula: CH3(CH2)3CONHNH2
Formula Weight: 116.16
Melting Point: 55-64°
Beilstein Reference: 1744747
Vzorec:
C₅H₁₂N₂O MW: 116,16 g/mol Bod tání: 55…64 °C Storage Temperature: Ambient |
MDL Number:
MFCD00025137 CAS číslo: 38291-82-6 EINECS: 253-864-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...