Stearic acid hydrazide ≥97%
Dodavatel: Thermo Fisher Scientific
|
Varování
|
Synonyms:
Stearicacidhydrazide
A18291.09
A18291.18
A18291.30
A18291.09EA
1350
CZK
A18291.09
A18291.18
A18291.30
Stearic acid hydrazide ≥97%
Stearic acid hydrazide
Purity: 97%
CAS No.: 4130-54-5
Molecular Formula: CH3(CH2)16CONHNH2
Formula Weight: 298.51
Melting Point: 114-116°
MDL No.: MFCD00014764
Vzorec:
C₁₈H₃₈N₂O Bod tání: 114…116 °C Storage Temperature: Ambient |
MDL Number:
MFCD00014764 CAS číslo: 4130-54-5 EINECS: 223-946-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...