Diisopropyl methylphosphonate 95%
Dodavatel: Thermo Fisher Scientific
|
Varování
|
A12904.06
A12904.14
A12904.22
A12904.06EA
1640
CZK
A12904.06
A12904.14
A12904.22
Diisopropyl methylphosphonate 95%
Diisopropyl methylphosphonate
Purity: 95%
CAS No.: 1445-75-6
Molecular Formula: C7H17O3P
Formula Weight: 180.18
Refractive Index: 1.4100
Boiling Point: 50-51°/1mm
MDL No.: MFCD00015022
Vzorec:
C₇H₁₇O₃P MW: 180.18 g/mol Bod varu: 50…51 °C (1 mmHg) Hustota: 0.976 Storage Temperature: Ambient |
MDL Number:
MFCD00015022 CAS číslo: 1445-75-6 EINECS: 215-896-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...