(±)-α-Bromophenylacetic acid, Sigma-Aldrich®
Dodavatel: Merck
Synonyms:
α-Bromophenylacetic Acid, DL-α-Bromophenylacetic acid
M-Clarity™ quality level = MQ100
B75859-25G
B75859-5G
SIALB75859-25GEA
4730
CZK
SIALB75859-25G
SIALB75859-5G
(±)-α-Bromophenylacetic acid, Sigma-Aldrich®
(±)-α-Bromophenylacetic acid
Vzorec:
C₈H₇BrO₂ MW: 215,0440 g/mol |
MDL Number:
MFCD00004206 CAS číslo: 4870-65-9 EINECS: 225-477-4 UN: 3261 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...