Tisk…

Bis(2-bromoethyl)ether, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: 1-Bromo-2-(2-bromoethoxy)ethane, 2-Bromoethyl ether

M-Clarity™ quality level = MQ100

382205-25ML 382205-5ML
SIAL382205-25MLEA 7590 CZK
SIAL382205-25ML SIAL382205-5ML
Bis(2-bromoethyl)ether, Sigma-Aldrich®
Bis(2-bromoethyl)ether
Vzorec: C₄H₈Br₂O
MW: 231.93 g/mol
MDL Number: MFCD00039196
CAS číslo: 5414-19-7
EINECS: 226-504-2
UN: 2810
ADR: 6.1 II

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR