Tisk…
Tetradecylphosphonic acid
Dodavatel: Cayman Chemical
Synonyms: 1-Tetradecylphosphonic acid, n-Tetradecylphosphonic acid, TDPA, N-tetradecylphosphonicacid
10007565-100 10007565-500 10007565-1
CAYM10007565-100EA 828 CZK
CAYM10007565-100 CAYM10007565-500 CAYM10007565-1
Tetradecylphosphonic acid
Tetradecylphosphonic acid
Vzorec: C₁₄H₃₁O₃P
MW: 278.37 g/mol
MDL Number: MFCD00015834
CAS číslo: 4671-75-4
EINECS: 225-118-1