Ethylene chlorophosphite ≥97%
Dodavatel: Thermo Scientific
|
Nebezpečí
|
A12724.14
A12724.22
A12724.36
A12724.14EA
3440
CZK
A12724.14
A12724.22
A12724.36
Ethylene chlorophosphite ≥97%
Ethylene chlorophosphite
Purity: 97%
CAS No.: 822-39-9
Molecular Formula: C2H4ClO2P
Formula Weight: 126.47
Storage Temperature: 2°C to 8°C
Flash Point: 86°(186°F)
Refractive Index: 1.4910
Boiling Point: 46-47°/15mm
Beilstein Reference: 506179
UN No.: 3265
MDL No.: MFCD00014511
Vzorec:
C₂H₄ClO₂P MW: 126,48 g/mol Bod varu: 46…47 °C (15 mmHg) Hustota: 1,415 Bod vzplanutí: 86 °C (186 °F) |
MDL Number:
MFCD00014511 CAS číslo: 822-39-9 EINECS: 212-499-4 UN: 3265 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...