Tisk…

3-Chloro-4-formylpyridine ≥98%

Dodavatel: Thermo Scientific
Varování

Synonyms: 3-Chloroisonicotinaldehyde, 3-Chloro-4-pyridinecarboxaldehyde, 3-Chloropyridine-4-carboxaldehyde

H55852.03 H55852.06
H55852.03EA 1470 CZK
H55852.03 H55852.06
3-Chloro-4-formylpyridine ≥98%
3-Chloro-4-formylpyridine

Purity: 98% CAS No.: 72990-37-5 Molecular Formula: C6H4ClNO Formula Weight: 141.56 Melting Point: 58-62° MDL No.: MFCD06200712

Vzorec: C₆H₄ClNO
MW: 141,56 g/mol
Bod tání: 58…62 °C
Storage Temperature: Ambient
MDL Number: MFCD06200712
CAS číslo: 72990-37-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR