2-Fluoro-4-nitroaniline 95%
Dodavatel: Thermo Scientific
|
Varování
|
H55174.06
H55174.14
H55174.06EA
1550
CZK
H55174.06
H55174.14
2-Fluoro-4-nitroaniline 95%
2-Fluoro-4-nitroaniline
Purity: 95%
CAS No.: 369-35-7
Molecular Formula: C6H5FN2O2
Formula Weight: 156.12
Melting Point: 122-130°
MDL No.: MFCD00034560
Vzorec:
C₆H₅FN₂O₂ MW: 156,12 g/mol Bod tání: 122…130 °C Storage Temperature: Ambient |
MDL Number:
MFCD00034560 CAS číslo: 369-35-7 EINECS: 206-719-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...