Tisk…

4-Amino-2-chloropyrimidine 98%

Dodavatel: Thermo Scientific
Nebezpečí

Synonyms: 2-Chlor-4-pyrimidinamin, 2-Chloro-4-pyrimidinamine, 2-Chloropyrimidin-4-amine, 2-Chloropyrimidin-4-ylamine

H55875.03 H55875.06
H55875.03EA 1680 CZK
H55875.03 H55875.06
4-Amino-2-chloropyrimidine 98%
4-Amino-2-chloropyrimidine

Purity: 98% CAS No.: 7461-50-9 Molecular Formula: C4H4ClN3 Formula Weight: 129.55 Melting Point: 197-205° MDL No.: MFCD00194051

Vzorec: C₄H₄ClN₃
MW: 129,55 g/mol
Bod tání: 197…205 °C
Storage Temperature: Ambient
MDL Number: MFCD00194051
CAS číslo: 7461-50-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR