4-Phenylaniline 98%
Dodavatel: Thermo Fisher Scientific
|
Nebezpečí
|
Synonyms:
4-Biphenylylamine, Biphenyl-4-ylamine, 4-Aminobiphenyl
H55424.03
H55424.06
H55424.14
H55424.03EA
893
CZK
H55424.03
H55424.06
H55424.14
4-Phenylaniline 98%
4-Phenylaniline
Purity: 98%
CAS No.: 92-67-1
Molecular Formula: C12H11N
Formula Weight: 169.23
Melting Point: 52-54°
Boiling Point: 191°/15mm
Merck Reference: 14,1235
UN No.: 3077
MDL No.: MFCD00007879
Vzorec:
C₁₂H₁₁N Bod varu: 191 °C (15 mmHg) Bod tání: 52…54 °C Hustota: 1,16 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00007879 CAS číslo: 92-67-1 EINECS: 202-177-1 UN: 3077 ADR: 9,III Merck Index: 14,1235 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...