1-Bromo-3,5-dimethoxybenzene ≥97%
Dodavatel: Thermo Fisher Scientific
H55312.06
H55312.14
H55312.06EA
1130
CZK
H55312.06
H55312.14
1-Bromo-3,5-dimethoxybenzene ≥97%
1-Bromo-3,5-dimethoxybenzene
Purity: 97%
CAS No.: 20469-65-2
Molecular Formula: C8H9BrO2
Formula Weight: 217.06
Melting Point: 62-66°
MDL No.: MFCD06200685
Vzorec:
C₈H₉BrO₂ Bod tání: 62…66 °C Storage Temperature: Ambient |
MDL Number:
MFCD06200685 CAS číslo: 20469-65-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...