Tisk…
Adapters, delivery, straight
Dodavatel: witeg Labortechnik
1.900.019 1.900.024 1.900.029
WITG1.900.019EA 537 CZK
WITG1.900.019 WITG1.900.024 WITG1.900.029
Adapters, delivery, straight
Komponenty pro destilaci Adaptéry destilační
Borosilicate glass 3.3.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes