Tisk…
Original Perfusor® syringes
Dodavatel: B. Braun
8728844F 8728828F 8728861F-06 8728852F-06 8728810F-06
613-0120EA 4890 CZK
613-0120 BRAU8728828F BRAU8728861F-06 BRAU8728852F-06 BRAU8728810F-06
Original Perfusor® syringes
Stříkačky Střílkačky zdravotnické
PP syringe with safe plunger backstop and minimal residual volume.
  • Oval grip plate for handling of the syringe and for prevention of rolling
  • Plunger stopper with double sealing ring for slow aspiration or infusion of drug solutions, made of synthetic material or natural latex-free
  • Centric Luer Lock cone for the attachment of a needle or infusion tubing