Tisk…

4,4'-Dimethyl-2,2'-bipyridyl, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: 4,4'-Dimethyl-2,2'-bipyridine

M-Clarity™ quality level = MQ200

245739-25G 245739-5G
SIAL245739-25GEA 15480 CZK
SIAL245739-25G SIAL245739-5G
4,4'-Dimethyl-2,2'-bipyridyl, Sigma-Aldrich®
4,4'-Dimethyl-2,2'-bipyridyl
Vzorec: C₁₂H₁₂N₂
MW: 184,24 g/mol
MDL Number: MFCD00006441
CAS číslo: 1134-35-6
EINECS: 214-483-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR