Tisk…

4,4'-Dimethyl-2,2'-bipyridyl ≥98%

Dodavatel: Thermo Fisher Scientific
Varování

Synonyms: 4,4'-Dimethyl-2,2'-bipyridine

B24936.03 B24936.06 B24936.14
B24936.03EA 1550 CZK
B24936.03 B24936.06 B24936.14
4,4'-Dimethyl-2,2'-bipyridyl ≥98%
4,4'-Dimethyl-2,2'-bipyridyl

Purity: 98% CAS No.: 1134-35-6 Molecular Formula: C12H12N2 Formula Weight: 184.24 Melting Point: 171-175° Beilstein Reference: 128995

Vzorec: C₁₂H₁₂N₂
Bod tání: 171…175 °C
Storage Temperature: Ambient
MDL Number: MFCD00006441
CAS číslo: 1134-35-6
EINECS: 214-483-2

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR