Tisk…

1,3-Bis(2,4,6-trimethylphenyl)imidazolium chloride, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: 1,3-Dimesityl-1H-imidazol-3-ium chloride, 1,3-Dimesitylimidazolium chloride

M-Clarity™ quality level = MQ200

574066-1G 574066-5G
SIAL574066-1GEA 2330 CZK
SIAL574066-1G SIAL574066-5G
1,3-Bis(2,4,6-trimethylphenyl)imidazolium chloride, Sigma-Aldrich®
1,3-Bis(2,4,6-trimethylphenyl)imidazolium chloride
Vzorec: C₂₁H₂₅ClN₂
MW: 340,89 g/mol
MDL Number: MFCD02684541
CAS číslo: 141556-45-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR