Tisk…

1,2-Bis(dicyclohexylphosphino)ethane, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ100

479500-1G 479500-5G
SIAL479500-1GEA 4700 CZK
SIAL479500-1G SIAL479500-5G
1,2-Bis(dicyclohexylphosphino)ethane, Sigma-Aldrich®
1,2-Bis(dicyclohexylphosphino)ethane
Vzorec: C₂₆H₄₈P₂
MW: 422,607 g/mol
MDL Number: MFCD00015521
CAS číslo: 23743-26-2
UN: 1325
ADR: 4.1,II

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR