Tisk…

6-Chloro-1,3-dimethyluracil ≥97%

Dodavatel: Thermo Scientific
Varování

Synonyms: 6-Chloro-1,3-dimethylpyrimidine-2,4(1H,3H)-dione, 1,3-Dimethyl-6-chlorouracil

A12755.03 A12755.06
A12755.03EA 495 CZK
A12755.03 A12755.06
6-Chloro-1,3-dimethyluracil ≥97%
6-Chloro-1,3-dimethyluracil

Purity: 97% CAS No.: 6972-27-6 Molecular Formula: C6H7ClN2O2 Formula Weight: 174.59 Storage Temperature: -30°C to -10°C Melting Point: 109-111° Beilstein Reference: 144393 MDL No.: MFCD00038066

Vzorec: C₆H₇ClN₂O₂
MW: 174,59 g/mol
Bod tání: 109…111 °C
MDL Number: MFCD00038066
CAS číslo: 6972-27-6
EINECS: 230-205-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR