Tisk…

Ethoxyquin, Sigma-Aldrich®

Dodavatel: Merck

E8260-250G E8260-100G
SIALE8260-250GEA 4250 CZK
SIALE8260-250G SIALE8260-100G
Ethoxyquin, Sigma-Aldrich®
Ethoxyquin
Vzorec: C₁₄H₁₉NO
MDL Number: MFCD00023883
CAS číslo: 91-53-2
EINECS: 202-075-7

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR