Tisk…

9,10-Bis(phenylethynyl)anthracene, Sigma-Aldrich®

Dodavatel: Merck

264199-1G 264199-5G
SIAL264199-1GEA 2990 CZK
SIAL264199-1G SIAL264199-5G
9,10-Bis(phenylethynyl)anthracene, Sigma-Aldrich®
9,10-Bis(phenylethynyl)anthracene
Vzorec: C₃₀H₁₈
MDL Number: MFCD00012050
CAS číslo: 10075-85-1
EINECS: 233-210-8

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR