Tisk…

Acridine orange (hemizinc salt)

Dodavatel: Thermo Fisher Scientific
Varování

Synonyms: 3,6-Bis(dimethylamino)acridine hydrochloride zinc chloride double salt, Euchrysine 3RX, Acridine orange hemi(Zinc chloride) salt, Acridine orange, Basic orange 14, 3,6-Bis(dimethylamino)acridinium chloride hemi(zinc chloride salt), Euchrysin GNY, N,N,N′,N′-Tetramethylacridine-3,6-diamine monohydrochloride compd. with zinc dichloride

L13159.06 L13159.14
L13159.06EA 807 CZK
L13159.06 L13159.14
Acridine orange (hemizinc salt)
Acridine orange (hemizinc salt)

CAS No.: 10127-02-3 Molecular Formula: C17H19N3.HCl.ZnCl2 Formula Weight: 438.10 Beilstein Reference: 3734978 MDL No.: MFCD00081043

Vzorec: C₁₇H₂₀ClN₃·HCl·½ZnCl₂
Storage Temperature: Ambient
MDL Number: MFCD00081043
CAS číslo: 10127-02-3
EINECS: 233-353-6

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR