Tisk…

(1S)-(-)-Camphanic acid, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: (1S,4R)-4,7,7-Trimethyl-3-oxo-2-oxabicyclo[2.2.1]heptane-1-carboxylic acid

M-Clarity™ quality level = MQ100

328227-1G 328227-5G
SIAL328227-1GEA 0 CZK
SIAL328227-1G SIAL328227-5G
(1S)-(-)-Camphanic acid, Sigma-Aldrich®
(1S)-(-)-Camphanic acid
Vzorec: C₁₀H₁₄O₄
MW: 198,22 g/mol
MDL Number: MFCD00044948
CAS číslo: 13429-83-9
EINECS: 236-552-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR