Tisk…
Amphotericin B 250 µl/ml in aqueous solution, Sigma-Aldrich®
Dodavatel: Merck
Synonyms: (1R,3S,5R,6R,9R,11R,15S,16R,17R,18S,19E,21E,23E,25E,27E,29E,31E,33R,35S,36R,37S)-33-(((2R,3S,4S,5S,6R)-4-Amino-3,5-dihydroxy-6-methyltetrahydro-2H-pyran-2-yl)oxy)-1,3,5,6,9,11,17,37-octahydroxy-15,16,18-trimethyl-13-oxo-14,39-dioxabicyclo[33.3.1]nonatriaconta-19,21,23,25,27,29,31-heptaene-36-carboxylic acid
M-Clarity™ quality level = MQ200
A2942-100ML A2942-20ML A2942-50ML
SIALA2942-100MLEA 5290 CZK
SIALA2942-100ML SIALA2942-20ML SIALA2942-50ML
Amphotericin B 250 µl/ml in aqueous solution, Sigma-Aldrich®
Amphotericin B
Vzorec: C₄₇H₇₃NO₁₇
MW: 924.08 g/mol
MDL Number: MFCD00877763
CAS číslo: 1397-89-3
EINECS: 215-742-2