Acriflavine, Sigma-Aldrich®
Dodavatel: Merck
|
Nebezpečí
|
M-Clarity™ quality level = MQ100
01673-10G
01673-50G
SIAL01673-10GEA
2700
CZK
SIAL01673-10G
SIAL01673-50G
Acriflavine, Sigma-Aldrich®
Acriflavine
Vzorec:
C₂₇H₂₅ClN₆ Storage Temperature: Ambient |
MDL Number:
MFCD00064307 CAS číslo: 8048-52-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...