Tisk…
DNA (Deoxyribonucleic acid) (from herring sperm), Sigma-Aldrich®
Dodavatel: Merck
D7290-1ML D7290-5X1ML
SIALD7290-1MLEA 1680 CZK
SIALD7290-1ML SIALD7290-5X1ML
DNA (Deoxyribonucleic acid) (from herring sperm), Sigma-Aldrich®
DNA (Deoxyribonucleic acid)
MDL Number: MFCD00130922
CAS číslo: 9007-49-2
EINECS: 309-566-6