Tisk…
DNA (Deoxyribonucleic acid) (from fish sperm), Sigma-Aldrich®
Dodavatel: Merck
M-Clarity™ quality level = MQ100
74782-100G 74782-10G
SIAL74782-100GEA 9170 CZK
SIAL74782-100G SIAL74782-10G
DNA (Deoxyribonucleic acid) (from fish sperm), Sigma-Aldrich®
DNA (Deoxyribonucleic acid)
MDL Number: MFCD00283430
CAS číslo: 9007-49-2
EINECS: 309-566-6