Tisk…

n-Undecyl-β-D-maltoside, Sigma-Aldrich®

Dodavatel: Merck

94206-250MG 94206-1G
SIAL94206-250MGEA 5090 CZK
SIAL94206-250MG SIAL94206-1G
n-Undecyl-β-D-maltoside, Sigma-Aldrich®
n-Undecyl-β-D-maltoside
Vzorec: C₂₃H₄₄O₁₁
CAS číslo: 170552-39-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR