Tisk…

Vanadium(III) chloride tetrahydrofuran complex (1:3), Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ100

395382-25G 395382-5G
SIAL395382-25GEA 9800 CZK
SIAL395382-25G SIAL395382-5G
Vanadium(III) chloride tetrahydrofuran complex (1:3), Sigma-Aldrich®
Vanadium(III) chloride tetrahydrofuran complex (1:3)
Vzorec: VCl₃·3C₄H₈O
MW: 373,62 g/mol
MDL Number: MFCD00145515
CAS číslo: 19559-06-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR