Tisk…

Trichlorotris(tetrahydrofuran)titanium(III) 97%

Dodavatel: Thermo Scientific
Nebezpečí

375160010 375160100
ACRO375160010EA 1560 CZK
ACRO375160010 ACRO375160100
Trichlorotris(tetrahydrofuran)titanium(III) 97%
Trichlorotris(tetrahydrofuran)titanium(III)
Vzorec: C₁₂H₂₄Cl₃O₃Ti
MW: 370,543 g/mol
Bod tání: >250 °C
Storage Temperature: Refrigerator
MDL Number: MFCD01631276
CAS číslo: 18039-90-2
UN: 2925
ADR: 4.1,II

Objednat

Výsledky specifikačního testu

Appearance Blue-green to blue Powder or crystals
Infrared spectrum Conforms
Titration Argentometric ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR