Tisk…

Tetrachlorobis(tetrahydrofuran)titanium(IV), Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ100

395404-25G 395404-5G
SIAL395404-25GEA 3980 CZK
SIAL395404-25G SIAL395404-5G
Tetrachlorobis(tetrahydrofuran)titanium(IV), Sigma-Aldrich®
Tetrachlorobis(tetrahydrofuran)titanium(IV)
Vzorec: C₄H₈Cl₄OTi
MW: 333,91 g/mol
MDL Number: MFCD00077884
CAS číslo: 31011-57-1
UN: 3261
ADR: 8,II

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR