(+)-Dibenzoyl-D-tartaric acid monohydrate ≥99%
Dodavatel: Thermo Fisher Scientific
|
Varování
|
Synonyms:
(2S,3S)-2,3-Bis(benzoyloxy)succinic acid hydrate
A16503.14
A16503.22
A16503.14EA
1320
CZK
A16503.14
A16503.22
(+)-Dibenzoyl-D-tartaric acid monohydrate ≥99%
(+)-Dibenzoyl-D-tartaric acid monohydrate
Purity: 99%
CAS No.: 80822-15-7
Molecular Formula: C18H14O8.H2O
Formula Weight: 376.33 (358.31anhy)
Melting Point: 87-91°
Beilstein Reference: 6121616
Vzorec:
C₁₈H₁₄O₈·1H₂O MW: 376.33 g/mol (358.31 anhy) Bod tání: 87…91 °C Storage Temperature: Ambient |
MDL Number:
MFCD00150722 CAS číslo: 80822-15-7 EINECS: 241-097-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...