(-)-Dibenzoyl-L-tartaric acid monohydrate, Sigma-Aldrich®
Dodavatel: Merck
Synonyms:
(2R,3R)-(-)-Di-O-benzoyl-tartaric acid monohydrate
33622-100G
33622-500G
SIAL33622-100GEA
1580
CZK
SIAL33622-100G
SIAL33622-500G
(-)-Dibenzoyl-L-tartaric acid monohydrate, Sigma-Aldrich®
(-)-Dibenzoyl-L-tartaric acid monohydrate
Vzorec:
C₁₈H₁₄O₈·1H₂O |
MDL Number:
MFCD00149119 CAS číslo: 62708-56-9 EINECS: 220-374-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...