Tisk…

Nystatin, Sigma-Aldrich®

Dodavatel: Merck
Varování

Synonyms: Fungicidin, Mycostatin

M-Clarity™ quality level = MQ200

N6261-25MU N6261-500KU N6261-5MU
SIALN6261-25MUEA 20000 CZK
SIALN6261-25MU SIALN6261-500KU SIALN6261-5MU
Nystatin, Sigma-Aldrich®
Nystatin
Vzorec: C₄₇H₇₅NO₁₇
MW: 926,09 g/mol
Storage Temperature: Freezer
MDL Number: MFCD25541651
CAS číslo: 1400-61-9
EINECS: 215-749-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR