Amphotericin B, Sigma-Aldrich®
Dodavatel: Merck
|
Varování
|
Synonyms:
(1R,3S,5R,6R,9R,11R,15S,16R,17R,18S,19E,21E,23E,25E,27E,29E,31E,33R,35S,36R,37S)-33-(((2R,3S,4S,5S,6R)-4-Amino-3,5-dihydroxy-6-methyltetrahydro-2H-pyran-2-yl)oxy)-1,3,5,6,9,11,17,37-octahydroxy-15,16,18-trimethyl-13-oxo-14,39-dioxabicyclo[33.3.1]nonatriaconta-19,21,23,25,27,29,31-heptaene-36-carboxylic acid
MQ300
A9528-100MG
A9528-1G
A9528-500MG
A9528-50MG
A9528-5G
SIALA9528-100MGEA
4190
CZK
SIALA9528-100MG
SIALA9528-1G
SIALA9528-500MG
SIALA9528-50MG
SIALA9528-5G
Amphotericin B, Sigma-Aldrich®
Amphotericin B
Vzorec:
C₄₇H₇₃NO₁₇ MW: 924.08 g/mol Storage Temperature: Freezer |
MDL Number:
MFCD00877763 CAS číslo: 1397-89-3 EINECS: 215-742-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...