Tisk…
2'-Deoxycytidine hydrochloride, Sigma-Aldrich®
Dodavatel: Merck
Synonyms: 4-Amino-1-((2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)pyrimidin-2(1H)-one hydrochloride
MQ200
D0776-1G D0776-250MG
SIALD0776-1GEA 3220 CZK
SIALD0776-1G SIALD0776-250MG
2'-Deoxycytidine hydrochloride, Sigma-Aldrich®
2'-Deoxycytidine hydrochloride
Vzorec: C₉H₁₄ClN₃O₄
MW: 263.68 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00012840
CAS číslo: 3992-42-5
EINECS: 223-639-9