2'-Deoxycytidine hydrochloride, Supelco®
Dodavatel: Merck
Synonyms:
4-Amino-1-((2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)pyrimidin-2(1H)-one hydrochloride
D8006-10G
D8006-1G
D8006-10GEA
17680
CZK
D8006-10G
D8006-1G
2'-Deoxycytidine hydrochloride, Supelco®
2'-Deoxycytidine hydrochloride
Vzorec:
C₉H₁₄ClN₃O₄ MW: 263.68 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00012840 CAS číslo: 3992-42-5 EINECS: 223-639-9 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...