Cytidine-5'-triphosphoric acid disodium salt (CTP-Na2) hydrate, Sigma-Aldrich®
Dodavatel: Merck
Synonyms:
5-CTP-Na2, CTP
30320-100MG
30320-500MG
SIAL30320-100MGEA
4050
CZK
SIAL30320-100MG
SIAL30320-500MG
Cytidine-5'-triphosphoric acid disodium salt (CTP-Na2) hydrate, Sigma-Aldrich®
Cytidine-5'-triphosphoric acid disodium salt (CTP-Na2) hydrate
Vzorec:
C₉H₁₄N₃Na₂O₁₄P₃·xH₂O MW: 527,12 g/mol (anhy) |
MDL Number:
MFCD00150809 CAS číslo: 652154-13-7 EINECS: 252-849-3 |
Learn more
![](/m-cz.vwr.com/cs_CZ/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...