Tisk…
Adenosine-5'-triphosphate disodium salt (ATP disodium salt) hydrate, Sigma-Aldrich®
Dodavatel: Merck
Synonyms: ATP disodium salt hydrate, 5′-ATP-Na2 ATP disodium salt hydrate, Adenosine 5'-Triphosphate Disodium Salt Hydrate, Adenosine-5'-triphosphate disodium salt hydrate, 5'-ATP 2Na Hydrate, ((2R,3S,4R,5R)-5-(6-Amino-9H-purin-9-yl)-3,4-dihydroxytetrahydrofuran-2-yl)methyl tetrahydrogen triphosphate disodium salt hydrate, ((2R,3S,4R,5R)-5-(6-Amino-9H-purin-9-yl)-3,4-dihydroxytetrahydrofuran-2-yl)methyl tetrahydrogen triphosphate, disodium salt xhydrate, Adenosine 5'-triphosphoric acid disodium hydrate
M-Clarity™ quality level = MQ200
A26209-10G A26209-1G A26209-5G
SIALA26209-10GEA 7490 CZK
SIALA26209-10G SIALA26209-1G SIALA26209-5G
Adenosine-5'-triphosphate disodium salt (ATP disodium salt) hydrate, Sigma-Aldrich®
Adenosine-5'-triphosphate disodium salt (ATP disodium salt) hydrate
Vzorec: C₁₀H₁₄N₅Na₂O₁₃P₃·xH₂O
MW: 551.14 g/mol (anhy)
MDL Number: MFCD00150755
CAS číslo: 34369-07-8
EINECS: 213-579-1