Tisk…

2-Fluoroadenosine, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: (2R,3R,4S,5R)-2-(6-Amino-2-fluoro-9H-purin-9-yl)-5-(hydroxymethyl)tetrahydrofuran-3,4-diol

M-Clarity™ quality level = MQ100

656402-1G 656402-250MG
SIAL656402-1GEA 16010 CZK
SIAL656402-1G SIAL656402-250MG
2-Fluoroadenosine, Sigma-Aldrich®
2-Fluoroadenosine
Vzorec: C₁₀H₁₂FN₅O₄
MW: 285.23 g/mol
MDL Number: MFCD00866394
CAS číslo: 146-78-1

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR