Tisk…

2'-Deoxyadenosine monohydrate, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: (2R,3S,5R)-5-(6-Amino-9H-purin-9-yl)-2-(hydroxymethyl)tetrahydrofuran-3-ol hydrate

M-Clarity™ quality level = MQ200

D8668-1G D8668-5G
SIALD8668-1GEA 4070 CZK
SIALD8668-1G SIALD8668-5G
2'-Deoxyadenosine monohydrate, Sigma-Aldrich®
2'-Deoxyadenosine monohydrate
Vzorec: C₁₀H₁₃N₅O₃·1H₂O
MW: 269.26 g/mol (251.24 anhy)
Storage Temperature: Ambient
MDL Number: MFCD00149364
CAS číslo: 16373-93-6
EINECS: 213-488-7

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR