Tisk…

2'-Deoxyadenosine monohydrate, Sigma-Aldrich®

Dodavatel: Merck

D7400-250MG D7400-500MG D7400-1G D7400-25G D7400-5G
SIALD7400-250MGEA 2020 CZK
SIALD7400-250MG SIALD7400-500MG SIALD7400-1G SIALD7400-25G SIALD7400-5G
2'-Deoxyadenosine monohydrate, Sigma-Aldrich®
Vzorec: C₁₀H₁₃N₅O₃·1H₂O
Storage Temperature: Ambient
MDL Number: MFCD00149364
CAS číslo: 16373-93-6
EINECS: 213-488-7

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR