Tisk…

2'-Deoxyadenosine monohydrate ≥98%, powder

Dodavatel: MP Biomedicals

Synonyms: (2R,3S,5R)-5-(6-Amino-9H-purin-9-yl)-2-(hydroxymethyl)tetrahydrofuran-3-ol hydrate

0210148125 0210148105
ICNA0210148125EA 31470 CZK
ICNA0210148125 ICNA0210148105
2'-Deoxyadenosine monohydrate ≥98%, powder
2'-Deoxyadenosine monohydrate

2′-Deoxyadenosine (2′-dAdo), a deoxyribonucleoside, may be used by some cells as an energy source under energy stress conditions and to affect cAMP levels. 2′-dAdo is used in comparison studies of the functions of adenosine analogues on various biological processes.


2′-Deoxyadenosine (2′-dAdo), a deoxyribonucleoside, may be used by some cells as an energy source under energy stress conditions and to affect cAMP levels. 2′-dAdo is used in comparison studies of the functions of adenosine analogues on various biological processes.


Soluble in water (50 mg/ml).

Vzorec: C₁₀H₁₃N₅O₃·1H₂O
MW: 269.26 g/mol (251.24 anhy)
Storage Temperature: Ambient
MDL Number: MFCD00149364
CAS číslo: 16373-93-6
EINECS: 213-488-7

Objednat

Výsledky specifikačního testu

Identity Test Passes
Purity ≥98%
FTIR Conforms to Standard
Melting Point 186 - 196 °C
UV/Vis: l max (water) 259 ± 2 nm
OD 250/OD 260 0,70 - 0,85
OD 280/OD 260 0,10 - 0,16


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR