Tisk…

Guanosine 5'-triphosphate trisodium salt hydrate, Sigma-Aldrich®

Dodavatel: Merck

G8877-100MG G8877-10MG G8877-250MG G8877-25MG G8877-1G
SIALG8877-100MGEA 9420 CZK
SIALG8877-100MG SIALG8877-10MG SIALG8877-250MG SIALG8877-25MG SIALG8877-1G
Guanosine 5'-triphosphate trisodium salt hydrate, Sigma-Aldrich®
Guanosine 5'-triphosphate trisodium salt hydrate
Vzorec: C₁₀H₁₃N₅Na₃O₁₄P₃·H₂O
Storage Temperature: Freezer
MDL Number: MFCD00077781
CAS číslo: 36051-31-7
EINECS: 252-847-2

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR