Tisk…

Guanosine-5'-monophosphate disodium salt hydrate, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ100

G8377-1G G8377-500MG G8377-100G G8377-25G
SIALG8377-1GEA 2370 CZK
SIALG8377-1G SIALG8377-500MG SIALG8377-100G SIALG8377-25G
Guanosine-5'-monophosphate disodium salt hydrate, Sigma-Aldrich®
Guanosine-5'-monophosphate disodium salt hydrate
Vzorec: C₁₀H₁₂N₅Na₂O₈P·H₂O
MW: 407.187 g/mol (anhy)
MDL Number: MFCD00151238
CAS číslo: 5550-12-9
EINECS: 226-914-1

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR