Tisk…

Guanosine-5'-monophosphate disodium salt hexahydrate

Dodavatel: MP Biomedicals

Synonyms: 5'-Guanylic acid disodium salt hexahydrate, GMP disodium salt hexahydrate

0210070205 0210070225
ICNA0210070205EA 4050 CZK
ICNA0210070205 ICNA0210070225
Guanosine-5'-monophosphate disodium salt hexahydrate
Guanosine-5'-monophosphate disodium salt hexahydrate
Vzorec: C₁₀H₁₂N₅Na₂O₈P·6H₂O
MW: 407,187 g/mol (anhy)
MDL Number: MFCD00151238
CAS číslo: 5550-12-9
EINECS: 226-914-1

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR